Learn More
MP Biomedicals MPQuanti™ Immunofluorescence Analyzer
Product Code. 30280601 Shop All MP Biomedicals Products
Change view
Click to view available options
Sample Type:
Blood
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Type Unit Size
30280601 Blood Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30280601 Supplier MP Biomedicals Supplier No. 07IMA001
  • ONLINE EXCLUSIVE PRICE
    18048.35 NOK
    17022.90 NOK / Pack of 1
    Save 1025.45 NOK
    5% Off
 Restricted Item
In Stock
MPQuanti™ is based on detection of fluorescence emitted during an immunoassay with antigen-antibody interaction.
  • Time-resolved fluorescence immunoassay analyzer is a rapid, quantitative instrument that tests quantitative items such as cardiac markers, inflammation markers, hormones, vitamin D and diabetes.
  • It is an automatic in-vitro diagnostic device that measures the concentration of analytes contained in blood, serum, and plasma samples.
  • The immunofluorescence analyzer should be used only by medical professionals to carry out in-vitro diagnostic.
  • It can be used in central laboratories, outpatient / emergency laboratories, clinical departments, and other medical service points such as community medical points and physical examination centers and can also be used in pre-hospital first aid ambulances.
  • CE-marked
  • Dedicated to time-resolved fluorescence immunoassay
  • Compatible with different sample type: whole blood, serum and plasma
  • Built-in printer
  • Data storage: more than 10,000 test records
  • Wide range of languages software
  • For professional in-vitro diagnostic use
  • The device allows you to test:
  • Cardiac markers: CK-MB, Troponin I, Myoglobin and D-Dimer.
  • Inflammation markers: C-Reactive Protein, Interleukin-6 and Procalcitonin.
  • Diabetes and Vitamin D: HBA1c and 25-Hydroxyvitamin D.
  • Hormones: B-HCG, LH, Progesterone, Prolactin, TSH, T3, T4, Free T3, Free T4.
Specifications
Type Accuracy Reader
Sample Type Blood
Memory 10,000 test records