missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ MPPE1 Recombinant Protein

Product Code. p-3720219
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16113903 10 μg 10µg
16123903 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16113903 Supplier Abnova™ Supplier No. H00065258P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH02877.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 65258
Molecular Weight (g/mol) 58.74
Name MPPE1 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen LLFCEFLIYYLAIFQCNWPEVKTTASDGEQTTREPVLKAMFLADTHLLGEFLGHWLDKLRREWQMERAFQTALWLLQPEVVFILGDIFDEGKWSTPEAWADDVERFQKMFRHPSHVQLKVVAGNHDIGFHYEMNTYKVERFEKVFSSERLFSWKGINFVMVNSVALNGDGCGICSETEAELIEVSHRLNCSRELLWWLQPRLVLSGHTHSACEVHHGGRVPELSVPSFSWRNRNNPSFIMGSITPTDYTLSKCYLPREDVVLIIYCGVVGFLVVLTLTHFGLLASPFLSGLNLLGKRKTR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Common Name MPPE1
Gene Symbol MPPE1
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.