Learn More
MP7, MedChemExpress
Product Code. 16438677
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16438677
Brand: Medchem Express HY14440
MedChemExpress MP7 (PDK1 inhibitor) is a phosphoinositide-dependent kinase-1 (PDK1) inhibitor.
MP7 (PDK1 inhibitor) is a phosphoinositide-dependent kinase-1 (PDK1) inhibitor.
Specifications
Chemical Name or Material MP7
CAS 1001409-50-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H22F2N4O4
Quantity 5 mg
Synonym PDK1 inhibitor
Solubility Information DMSO : ≥ 100 mg/mL (193.61 mM)
SMILES O=C(C1=CC=CN(CC2=CC=C(F)C(F)=C2)C1=O)N[C@H](C3=CC=CC=C3)COC4=CC=C(N5)C(NC5=O)=C4
Molecular Weight (g/mol) 516.5
Formula Weight 516.5
Percent Purity 99.83%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.