Learn More
Plum™ MP2000 Dispenser
Product Code. 15771137
Change view
Click to view available options
Width (Metric):
130 mm
230 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
15771137 230 mm Each
15761137 130 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15771137 Supplier Plum™ Supplier No. P4252
  • ONLINE EXCLUSIVE PRICE
    1090.00 NOK
    788.15 NOK / Pack of 1
    Save 301.85 NOK
    27% Off
Estimated Shipment: 21-01-2027
Flexible dispenser system for bag-in-box products (soap/ hand cleaning/ hand lotion). Well-suited for production plants, repair shops, food processing, technical colleges and graphical industries.
  • Hygienic and reliable dispenser design
  • Easily customised to hand hygiene requirements in the individual workplace
  • MP2000 products are supplied in airtight packages for prevention of bacteria growth
  • Easily mounted, cleaned and refilled
  • Specifications
    Depth (Metric) 110 mm
    Width (Metric) 230 mm
    Height (Metric) 300 mm
    No. of Compartments 2