Learn More
MP Biomedicals™ MP EcoChrom Polyamide (for TLC)
Product Code. 30058482 Shop All MP Biomedicals Products
Change view
Click to view available options
:
Particle Size: 5 to 20 μm
Product Code.
30058482 Particle Size: 5 to 20 μm
1 options
Product Code. 30058482
Brand: MP Biomedicals™ 209603
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
MP EcoChrom Polyamide (for TLC)
  • Usage Statement: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 5 to 20 μm
Technique Chromatography
Type Polyamide
Material Polyamide
For Use With (Application) TLC