missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Val-Gly-Ser-Glu

Product Code. 11403941 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403941 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11403941 Supplier MP Biomedicals™ Supplier No. 0215277601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Content And Storage 0°C
Format Powder
Molecular Weight (g/mol) 390.392
Product Type Val-Gly-Ser-Glu
Quantity 1 mg
Sequence Val-Gly-Ser-Glu
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.