Learn More
MP Biomedicals™ TeenPrep™ Lysing Matrix Z, 15 mL Tubes
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 x 15 mL Tubes
5 x 15 mL Tubes
50 x 15 mL Tubes
Unit Size:
Pack of 25
Pack of 5
Pack of 50
Description
Lysing Matrix Z tubes can be used with FastPrep instruments or similar bead beaters. They are ideal for lysis of animal and plant tissue and whole insects. Lysing Matrix Z is an alternative to the very aggressive Lysing Matrix A for applications such as RNA and protein isolation from very tough tissue.
- Tube size: 15 mL
- Samples types: Animal tissue, plant tissue, tough stems, roots, seeds, whole insects
- Matrix composition: 2 mm diameter yttria-stabilized zirconium oxide beads
Specifications
Specifications
| Bead Type | Yttria-stabilized Zirconium Oxide |
| Sample Type | Animal Tissues, Plants, Insects |
| For Use With (Equipment) | FastPrep instrument or other bead beater |
| For Use With (Application) | Nucleic Acid Isolation and Purification, Homogenization |
| Matrix | Lysing Matrix Z |
| Quantity | 50 x 15 mL Tubes |