Learn More
MP Biomedicals™ TallPrep™ Lysing Matrix Y, 4.5 mL Tubes
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 x 4.5 mL
50 x 4.5 mL
100 x 4.5 mL
Unit Size:
Pack of 100
Pack of 25
Pack of 50
Description
Lysing Matrix Y tubes can be used with FastPrep instruments or similar bead beaters. The yttria-stabilized zirconium oxide beads have a high density and hardness for cracking tough cell walls, yet a smooth surface for minimizing shearing.
- Tube size: 4.5 mL
- Samples types: Gram (+/-) bacteria, fungal tissue and spores, yeast, molds, filamentous algae and biofilms
- Matrix composition: 0.5 mm diameter yttria-stabilized zirconium oxide beads
Especificaciones
Especificaciones
| Bead Type | Yttria-stabilized Zirconium Oxide |
| Sample Type | Yeast, Fungi, Algae |
| For Use With (Equipment) | FastPrep instruments or other bead beaters |
| For Use With (Application) | Nucleic Acid Isolation and Purification, Homogenization |
| Includes | 0.5mm dia. Yttria-Stabilized Zirconium Oxide beads |
| Matrix | Lysing Matrix Y |
| Quantity | 100 x 4.5 mL |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido