missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ready to Use 5% 7X™ Solutions

Product Code. 11481495 Shop All MP Biomedicals Products
Change view
Click to view available options
Description:
7X-O-Matic; 1 gal.
7X; 1 gal.
ES 7X; 1 gal.
Unit Size:
Each
Product Code. Description unitSize
11481495 7X; 1 gal. Each
11491495 ES 7X; 1 gal. Each
11401505 7X-O-Matic; 1 gal. Each
3 options
This item is not returnable. View return policy

Product Code. 11481495

Brand: MP Biomedicals™ 097668093

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A 1 gallon Ready to Use 5% solution of 7X that includes a hand-pump for convenient dispensing

  • Ideal for glass and plastic labware
  • Contain powerful sequestering reagents to prevent deposits that could harm equipment or critical experimental work, and they will not etch delicate glassware or plastic labware
  • Environmentally friendly, phosphate-free ES formulation is completely soluble in water at any concentration and drains completely from glassware in seconds

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Concentration 7X
Content And Storage 15°C to 30°C
Description 7X; 1 gal.
For Use With (Application) For Glass, Plastic, benchtop equipment and tubing
Volume (English) 1 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.