Description
- Ideal for glass and plastic labware
- Contain powerful sequestering reagents to prevent deposits that could harm equipment or critical experimental work, and they will not etch delicate glassware or plastic labware
- Environmentally friendly, phosphate-free ES formulation is completely soluble in water at any concentration and drains completely from glassware in seconds
Specifications
Specifications
| Form | Liquid |
| Concentration | 7X |
| Content And Storage | 15°C to 30°C |
| Description | 7X-O-Matic; 1 gal. |
| For Use With (Application) | For Glass, Plastic, benchtop equipment and tubing |
| Volume (English) | 1 gal. |