missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Concentration | ≥200 U/mg |
| Color | Clear |
| pH | 7.5 |
| For Use With (Application) | Used as a coupling Enzyme in conjunction with Lactic Dehydrogenase in quantifying ADP and the activity of Enzymes that catalyze the formation of ADP |
| Content And Storage | -20°C |
| Source | Rabbit muscle |
| Quantity | 5000 U |
| Product Type | Pyruvate Kinase |
| Molecular Weight (g/mol) | 237000 |
| Inhibitors | Activity is inhibited by Ca2+, Fluorophosphate inhibits the Enzyme reaction competitively with respect to PEP , ATP inhibits the reaction by removal of Mg2+ from the Substrate MgADP- |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?