missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Poly-L-Lysine Hydrobromide Attachment Factors: ≥70,000 MW

Product Code. 11450812 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Sterility:
Non-sterile
Sterile
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 11450812

Brand: MP Biomedicals™ 0219454405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Facilitate attachment, adherence, and spreading in the culture of adherent cells

Available nonsterile or gamma-irradiated.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Product Type Poly-L-Lysine
Sterility Sterile
Quantity 5 mg
Source Synthetic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.