missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Poly-L-Lysine Hydrobromide Attachment Factors: ≥70,000 MW
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Sterility:
Non-sterile
Sterile
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
Available nonsterile or gamma-irradiated.
Specifications
Specifications
| Content And Storage | -20°C |
| Product Type | Poly-L-Lysine |
| Sterility | Non-sterile |
| Quantity | 25 mg |
| Source | Synthetic |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction