missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Pepstatin A

Product Code. 11415332 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
100 mg
250 mg
Unit Size:
100mg
10mg
1mg
250mg
25mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415332 100 mg 100mg
11485322 5 mg 5mg
11405332 25 mg 25mg
11495322 10 mg 10mg
11475322 1 mg 1mg
11425332 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11415332 Supplier MP Biomedicals™ Supplier No. 0219536880

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic inhibitor for pepsin, renin, cathepsin D, and other acid proteases.

Pepstatin A, [26305-03-3]; Isoval-Val-Val-Sta-Ala-Sta

C34H63N5O9, F.W. 685.9; Melting Point: 235-237°C

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Enzyme Assay, Inhibition Assay, Molecular Biology, Protein Purification, Sample Preparation
Molecular Weight (g/mol) 685.91
Name Pepstatin A
Quantity 100 mg
Storage Requirements 0°C
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.