Learn More
MP Biomedicals™ Min. Ess. Med. Eagle, Alpha Modification
Product Code. 11480275 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480275 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11480275 Supplier MP Biomedicals™ Supplier No. 091031122
  • ONLINE EXCLUSIVE PRICE
    716.00 NOK
    681.19 NOK / 10 L
    Save 34.81 NOK
    4% Off
 Restricted Item
Estimated Shipment: 22-12-2026
Alpha Modification of Eagle's Medium (AMEM), powder, with L-glutamine, without deoxyribosides, ribosides, sodium bicarbonate
Specifications
Product Type Alpha MEM
Without Additives No Deoxyribonucleosides, No Ribonucleosides, No Sodium Bicarbonate
Quantity 10 L
pH 6.5 to 7.5
Content And Storage 0° to 5°C