missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Hemocyanin

Product Code. 11463771 Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
This item is not returnable. View return policy

Product Code. 11463771

Brand: MP Biomedicals™ 0215123350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Derived from Megathura crenulata

Hemocyanins are copper-containing oxygen transport proteins found in the hemolymph of many invertebrates. It is dissolved in the hemolymph; not found in blood cells. They are divided into 2 main groups, arthropodan (such as horseshoe crab) and molluscan (such as keyhole limpet). These have structurally similar oxygen-binding sites, which are similar to the oxygen-binding sites of tyrosinases, but their quaternary structures are arranged differently.

  • Format: Gray lyophilized powder

Immunology

TRUSTED_SUSTAINABILITY

Specifications

Formulation 5mM BES Buffer, pH 6.5 and 10mM MgSO4
Molecular Weight (g/mol) 3000000 to 7500000
Name Hemocyanin
Quantity 50 mg
Source Keyhole Limpet
Storage Requirements 4°C
Form Lyophilized
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.