missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ GS96 Media, Powdered
Shop All MP Biomedicals Products
Click to view available options
Quantity:
227 g
1000 g
Unit Size:
1kg
227g
Description
GS96 Broth is formulated to grow host cells to the highest density possible without aeration, and to produce high yields of plasmid DNA from those cultures. Perfect for growing bacterial cells in 96-well and 384-well plates, GS96 media can be used in any situation in which aeration of the culture is difficult or absent. GS96 broth is used at the front end of high-throughput automated procedures, and growth can be increased even further by the addition of 0.05% glycerol.
- Formulated to grow host cells to the highest density possible without aeration, and to produce high yields of plasmid DNA from those cultures
- Perfect for growing bacterial cells in 96- and 384-well plates
- Can be used in any situation in which aeration of the culture is difficult or absent
- Used at the front end of high-throughput automated procedures
- Growth can be increased even further by the addition of 0.05% glycerol
- Contents/L: proprietary
- Use 50g/L
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Form | Powder |
| Grade | Molecular biology |
| Product Type | Microbiology Media |
| Formulation | Yeast extract, Peptones, Vitamins, Trace Elements, Carbon and Nitrogen Sources, Small quantities of biological Buffers have been added for proper pH adjustment |
| For Use With (Application) | To grow host cells to the highest density possible without aeration and to produce high yields of plasmid DNA from those cultures |
| Quantity | 227 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction