missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ β- Galactosidase from Escherichia coli
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kU
10 kU
5 kU
Unit Size:
10ku
1ku
5ku
Description
- CAS Number: 9031-11-2
- EC Number: 232-864-1
- Format: Pale gray fuffy powder
- Specific activity: ≥50U/mg
- Isoelectric point: 4.61
- Optimum pH: 7 to 7.5
Used for structural investigation of carbohydrate, determination of lactose (foodstuff analysis) and as an enzyme label for enzyme immunoassay.
Specifications
Specifications
| Concentration | ≥50 U/mg |
| Color | Pale Gray |
| pH | 7 to 7.5 |
| For Use With (Application) | Structural investigation of Carbohydrate, the determination of Lactose (food stuff analysis) and as an Enzyme label for Enzyme immunoassay |
| Content And Storage | 4°C |
| Source | E. coli |
| Quantity | 5 kU |
| Product Type | beta-Galactosidase |
| Molecular Weight (g/mol) | 540000 |
| Inhibitors | p-Chloromercuribenzoate, lodoacetamide, Heavy Metal Ions (Zn++, Fe++, Zn++, Cd++, Cu++, Pb++, Ag+, Hg++), Ionic Detergents SDS, DAC |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction