Learn More
MP Biomedicals™ Screw Caps
Product Code. p-4940393 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1000 Pk.
500 Pk.
Color:
Blue
Green
Orange
Purple
Red
Unit Size:
Pack of 1000
Pack of 500
Product Code. Quantity Color unitSize
11472340 500 Pk. Red Pack of 500
11402340 500 Pk. Orange Pack of 500
11402350 500 Pk. Green Pack of 500
11442340 500 Pk. Blue Pack of 500
11422340 500 Pk. Purple Pack of 500
11412340 1000 Pk. Orange Pack of 1000
12733667 1000 Pk. Blue Pack of 1000
7 options
Product Code. 11472340
Brand: MP Biomedicals™ 115066002
These 2 mL Screw Caps are for tubes to be used on FastPrep™ instruments or any bead beater.
  • Screw Caps with O-rings fit 2 mL screw cap tubes and are ideal for high-speed sample preparation instruments.
  • Screw Caps fit Empty Fastprep™ Tubes and are used in Fastprep systems, to hold sample for Homogenization.
  • Available separately for those who wish to fill their own Lysing Matrix Tubes.
  • Adapter Compatibility: ConeFlex Legacy (allows all FP24 adapters on FP96), CoolPrep (24x2 mL), HiPrep (48x2 mL), QuickFlex (96x2 mL), QuickPrep (24x2 mL), QuickPrep All Metal (24x2 mL)
  • Screw Caps can also be found in the FastDNA and FastDNA SPIN Kits.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 2 mL
Color Red
Density 0.946 g/cm3
Sample Type All
For Use With (Equipment) 2 mL Lysing Matrix C Tubes
For Use With (Application) Nucleic Acid Isolation, Cell Culture, Proteomics, Purification and Detection, Protein Purification, DNA Purification, RNA Purification, Plant Tissue Processing, Tissue Analysis, Soil Analysis, Bacterial Culture, Yeast Culture, Fungal Tissue Analysis, Microbiome Studies
Melting Point 320°F (160°C)
Product Type Screw Cap
Quantity 500 Pk.
Remarks Lysing Matrix C tubes have red caps and are found in the Fast RNA Pro Red Kit for isolation of total RNA from yeast and fungi.
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.