Learn More
MP Biomedicals™ EcoChrome™ Silica Plates
Shop All MP Biomedicals Products
Click to view available options
Unit Size:
Pack of 2025
Specifications
Specifications
| Type | EcoChrome Silica Plate |
| For Use With (Application) | Chromatography, Chemical Synthesis, Quantitative Densitometric UV/Vis Evaluation |
| Storage Requirements | 15°C to 30°C |
| Dimensions (L x W x H) | 20 x 20 x 0.25 cm |