Learn More
MP Biomedicals™ Earle's Balanced Salts, 10X EBS Liq., Modified
Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| Chemical Name or Material | Earle's Balanced Salt |
| Color | Orange |
| Concentration | 10 X |
| pH | 4.44 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Formulation | Calcium Chloride Dihydrate 2649mg/L, Magnesium Sulfate 977mg/L, Potassium Chloride 4000mg/L, Sodium Chloride 68000mg/L, Sodium Phosphate Monobasic Monohydrate 1400mg/L, Dextrose 10000mg/L, Phenol Red Sodium Salt 170mg/L |