missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Dispase

Product Code. 11466791 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 U
5 U
Unit Size:
25 units
5 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466791 5 U 5 units
11476791 25 U 25 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466791 Supplier MP Biomedicals™ Supplier No. 0219502205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

From Bacillus polymyxa

Microbial metalloenzyme used in tissue dissociation (primary cell cultures) and subcultivation (secondary cell cultures). Extremely stable. Activity: 6U/mg. Unit Definition: One unit is the amount of enzyme which liberates 1.0μmole of Tyr per minute at 37°C, pH 7.5. MW: 35.9kDa. Lyophilized.

Ref.: Akimoto, Y., et al., J. Histochem. Cytochem., 40: 1129 (1992).

TRUSTED_SUSTAINABILITY

Specifications

Color Tan
pH 6 to 8.5
For Use With (Application) Protein sequencing
Content And Storage 4°C
Source Bacillus polymyxa
Quantity 5 U
Product Type Dispase
Molecular Weight (g/mol) 35.9
Inhibitors EDTA, EGTA, Hg2+, other heavy Metals
Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.