missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Diastase
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
Diastase, [9000-92-4]
- Activity: 1000 Lintner u/g
- Melting Point: 175°C (dec)
Specifications
Specifications
| Concentration | 1000 U/g |
| Color | Off-White |
| For Use With (Application) | Digests 50g of Starch in <30 min. |
| Content And Storage | 4°C |
| Source | Malt |
| Quantity | 100 g |
| Product Type | Diastase |
| Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction