Description
- Preweighed reagents in tablet form offer convenience and efficiency.
- Tablets both reduce the risk of exposure to hazardous dust and eliminate cumbersome weighing and waste.
- Simply drop into the appropriate amount of distilled water, dissolve, and proceed.
- 100 tablets/Pk.
- Dissolve one tablet in 10 mL deionized water. Use liquid substrate within one hour. Immediately before use, add hydrogen peroxide to 0.1% (v/v).
Specifications
Specifications
| Concentration | 5 mg of DAB per Tablet |
| For Use With (Application) | Immunohistology, immunoblotting |
| pH | 4.94 |
| Quantity | 100 x 5 mg |
| Form | Tablet |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.