missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CellMaxx™ Bovine Albumin, Low IgG, Stem Cell Culture
Lipid-rich bovine serum albumin, suitable for stem cell culture and media formulation
POA
Specifications
For Use With (Application) | ELISA, Mammalian Cell Culture, Western Blot |
---|---|
Formulation | Lipid-rich |
Molecular Weight (g/mol) | 66 |
Name | CellMaxx™ Bovine Albumin, Low IgG |
pH Range | 6.5 to 7.5 |
Description
Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells; IgG removed to very low levels. Lipid-rich formulation can be substituted for Albumax I™.
Physical characteristics:
- Form: Off white to brown powder
- Purity: ≥98% (w/w)
- Total Protein: ≥96%
- IgG 0.005% w/w
- pH (4%): 6.5 to 7.5
- Solubility: Dissolves in 20 minutes at 20° to 25°C
- Moisture: ≤5% w/w
- Endotoxin: ≤1EU/mg
- Mycoplasma: Negative
- Bioburden: ≤100cfu/g
- Virus (9CFR): Negative
- Reported: Lipid content
Specifications
ELISA, Mammalian Cell Culture, Western Blot | |
66 | |
6.5 to 7.5 | |
Bovine | |
Powder |
Lipid-rich | |
CellMaxx™ Bovine Albumin, Low IgG | |
Chromatography | |
4°C |