missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Cell Preparation Grade Collagenase from Clostridium histolyticum

Product Code. 11412082 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 x 1 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412082 5 x 1 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11412082 Supplier MP Biomedicals™ Supplier No. 0215070505

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prepared to contain lower tryptic activity to limit damage to membrane proteins and receptors but with normal to above normal collagenase activity

The collagenase product is a mixture of enzymes secreted by C. histolyticum. The main components are two collagenases, clostripain, and a neutral protease. The synergistic action of these enzymes degrade collagen and other intracellular material. The action of both collagenase enzymes and the neutral protease is necessary for effective release of cells from tissue. Various types of collagen are the natural substrates for collagenase.

TRUSTED_SUSTAINABILITY

Specifications

Color Brown
Content And Storage Store at 4°C. Protect from moisture.
Quantity 5 x 1 g
Product Type Collagenase
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.