missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Bacteriological Agar

Product Code. 11416501 Shop All MP Biomedicals Products
Change view
Click to view available options
:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11416501 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11416501 Supplier MP Biomedicals™ Supplier No. 0219402183

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specially purified for use in preparing solid culture media for microbiological and bacterilogical applications

Naturally occurring impurities have been reduced to a minimum

  • CAS: 9002-18-0
  • EC No. 232-658-1

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Form Powder
Grade Bacteriological
Product Type Agar
Formulation Agarose 70% and Agaropectin 30%
For Use With (Application) Used for preparing solid culture media for microbiological and bacteriological application, antistaling agent, gelling agent
Includes 2U of Agarose and Agaropectin
Quantity 250 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.