missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Spectinomycin, dihydrochloride pentahydrate, PhytoPure™

Product Code. p-4890780 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
17610917 1 g 1g
17620917 5 g 5g
11470802 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 17610917

Brand: MP Biomedicals™ 0219454001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An aminocyclitol antibiotic, closely related to the aminoglycosides, it inhibits protein synthesis (elongation) by interfering with peptidyl tRNA translocation. It is active against gram-negative bacteria and is used as a selection agent for transformed plant cells that contain the selectable marker gene Spcr.

 

  • Format: Crystalline Powder, Non-sterile
  • Synonyms: U18409AE; Actinospectacin; M141; Trobicin 

Specinomycin is used in cell culture applications, It is active against gram-negative bacteria.It has been shown to have a wide variety of uses, including treating acute gonorrheal urethritis and cervictis, to mark cell layers to monitor cell fate during leaf development, as a selection marker in plant related transformation systems for plant cells containing the marker gene Spcr, to study respiratory tract infections of cattle caused by Pasteurella multocida and Mannheimia haemolytica, and to generate plants deficient for the plastid-encoded RNA polymerase on MS-agar media. Spectinomycin also has use in amplification of low copy number plasmid carrying replicons.

All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.

TRUSTED_SUSTAINABILITY

Spezifikation

CAS 22189-32-8
Content And Storage 4°C
Form Crystalline powder
Product Type Antibiotic
Quantity 1 g
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.