Learn More
MP Biomedicals™ Angiotensin-Converting Enzyme Substrate
Product Code. 11473901 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. 11473901
Brand: MP Biomedicals™ 0215274790
Free acid
Hippuryl-His-Leu (Benzoyl-Gly-His-Leu)
  • A fluorogenic substrate for angiotensin-converting enzyme
TRUSTED_SUSTAINABILITY
Specifications
Color Off-White
For Use With (Application) Continuous spectrophortometric assays of Angiotensin Converting Enzyme
Content And Storage -20°C
Quantity 500 mg
Product Type Angiotensin-converting Enzyme Substrate
Molecular Weight (g/mol) 399.4
Form Powder