missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Acid Phosphatase, Wheat Germ

Product Code. 11447312 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447312 500 mg 500mg
11457312 1 g 1g
11467312 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447312 Supplier MP Biomedicals™ Supplier No. 0215387690

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

  • Activity 20units/mg

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20 U/mg
Color Brown
Purity 90%
pH 4 to 7
For Use With (Application) Catalyzes the hydrolysis of Monoesters and Anhydrides of Phosphoric Acid
Content And Storage -20°C
Source Wheat germ
Quantity 500 mg
Product Type Acid Phosphatase
Molecular Weight (g/mol) 58
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.