Learn More
MP BIOMEDICALS MP Alumina B
Shop All MP Biomedicals Products
Click to view available options
:
Alumina B
Description
32 to 63 μm, active, basic pH alumina B for chromatography
Specifications
Specifications
| Particle Size | 32 to 63 μm |
| Technique | Chromatography |
| Type | Alumina B |
| Material | Alumina B |