Learn More
Moxifloxacin, MedChemExpress
Product Code. 16447916
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16447916 100 mg 100mg
16411946 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16447916 Supplier Medchem Express Supplier No. HY66011A
MedChemExpress Moxifloxacin is an orally active 8-methoxyquinolone antimicrobial for use in the treatment of acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia.
Moxifloxacin is an orally active 8-methoxyquinolone antimicrobial for use in the treatment of acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia.
Specifications
Chemical Name or Material Moxifloxacin
Physical Form Solid
CAS 151096-09-2
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H24FN3O4
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (249.11 mM; Need ultrasonic)
SMILES O=C(C1=CN(C2CC2)C3=C(C=C(F)C(N(C4)C[C@]5([H])[C@@]4([H])CCCN5)=C3OC)C1=O)O
Molecular Weight (g/mol) 401.43
Formula Weight 401.43
Percent Purity 99.48%
Grade Research
Show More Show Less
Research purposes only
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.