missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOXD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11545-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
MOXD1 | |
Polyclonal | |
Unconjugated | |
MOXD1 | |
Monooxygenase X, monooxygenase, DBH like 1, MOX, MOXD1, PRO5780 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26002, 294119, 59012 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6UVY6, Q9CXI3 | |
Moxd1 | |
MOXD1 Fusion Protein Ag2060 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |