Learn More
Moxalactam sodium salt, MedChemExpress
Product Code. 16492276
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
16492276 500 mg 500mg
1 options
Product Code. 16492276
Brand: Medchem Express HYB1484
MedChemExpress Moxalactam (Latamoxef) sodium salt is a synthetic oxa-β-lactam antibiotic. Moxalactam sodium salt has a broad spectrum of activity against Gram-positive and Gram-negative aerobic and anaerobic bacteria. Moxalactam sodium salt inhibits production of β-lactamases.
Moxalactam (Latamoxef) sodium salt is a synthetic oxa-β-lactam antibiotic. Moxalactam sodium salt has a broad spectrum of activity against Gram-positive and Gram-negative aerobic and anaerobic bacteria. Moxalactam sodium salt inhibits production of β-lactamases.
Specifications
Chemical Name or Material Moxalactam sodium salt
Physical Form Powder
CAS 64953-12-4
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C20H18N6Na2O9S
For Use With (Application) COVID-19-immunoregulation
Synonym Latamoxef sodiumLamoxactam sodiumLY-127935 sodium
Solubility Information H2O : 100 mg/mL (177.17 mM; Need ultrasonic) ∣DMSO : 62.5 mg/mL (110.73 mM; Need ultrasonic)
SMILES O=C(C(N12)=C(CSC3=NN=NN3C)CO[C@]2([H])[C@](OC)(NC([C@H](C(O[Na])=O)C4=CC=C(O)C=C4)=O)C1=O)O[Na]
Molecular Weight (g/mol) 564.44
Formula Weight 564.44
Percent Purity 95.0%
Grade Research