missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MOV10 Monoclonal antibody specifically detects MOV10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
Specifications
| Antigen | MOV10 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | SR1069 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Formulation | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gene Alias | DKFZp667O1423, EC 3.6.1, EC 3.6.4.13, FLJ32791, KIAA1631mouse) homolog, Mov10, Moloney leukemia virus 10, homolog (mouse) |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human MOV10 (Uniprot #: Q9HCE1) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?