missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TREM-1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 7 publications
Brand: R&D Systems MAB1187-100
This item is not returnable.
View return policy
Description
TREM1 Monoclonal specifically detects TREM1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Agonist Activity, Immunocytochemistry, CyTOF-ready.
Specifications
| TREM1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD354, CD354 antigen, TREM1, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54210 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Flow Cytometry, Agonist Activity, Immunocytochemistry, CyTOF | |
| 174031 | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Agonist Activity, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Q9JKE2 | |
| TREM1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TREM-1 Ala21-Ser202 Accession # Q9JKE2 | |
| 100 μg | |
| Primary | |
| Detects mouse TREM-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse TREM-2B, recombinant human (rh) TREM-1, or rhTREM-2 is observed. | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction