missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse TPO (Thrombopoietin), AOF Recombinant Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Description
Thrombopoietin (TPO, THPO, MPL ligand, megakaryocyte colony-stimulating factor) is responsible for the generation of platelets through its action on hematopoietic cells committed to the megakaryocyte lineage. Binding of thrombopoietin to its receptor, MPL (CD110), initiates a signaling cascade via JAK/STAT, MAPK, and PI3K/AKT signaling pathways that leads to the proliferation and differentiation of megakaryocytes and, ultimately, to the production of platelets (thrombopoiesis). Thrombopoietin is constitutively produced by the liver and has a predicted molecular weight of 38 kDa.
- Purity: >95% by SDS-Page
- Endotoxin Concentration: AX41 ≤EUs/μg
- Protein Length: 175 aa.
- Biological Activity: <5 ng/mL
- Molecular Weight: 18.7 kDa
- Animal Origin Free: Yes
Specifications
Specifications
| Accession Number | P40226 |
| For Use With (Application) | Control, ELISA, Functional Assay, Gel Shift, Western Blot |
| Formulation | 10 mM sodium phosphate with no preservative |
| Gene ID (Entrez) | 21832 |
| Molecular Weight (g/mol) | 18.7 kDa |
| Name | Mouse Thrombopoietin (TPO), AOF |
| Quantity | 1 mg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction