missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TCF7/TCF1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB8224V-100UG
This item is not returnable.
View return policy
Description
TCF7/TCF1 Monoclonal specifically detects TCF7/TCF1 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| TCF7/TCF1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q00417 | |
| TCF7 | |
| E. coli-derived recombinant mouse TCF7/TCF1 Gly61-Gly116 Accession # Q00417 | |
| 100 μg | |
| Primary | |
| Detects mouse TCF7/TCF1 in ELISA and Western Blot. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 812145 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| FLJ36364, MGC47735, T-cell factor 1, T-cell specific, TCF1, TCF-7, transcription factor 7 (T-cell specific, HMG-box) | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6932 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction