Learn More
STEMCELL Technologies Mouse Recombinant IL-10
Product Code. 17109521
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Product Code. Quantity unitSize
17109521 50 μg 50µg
17199511 10 μg 10µg
2 options
Product Code. 17109521
Brand: STEMCELL Technologies undefined
Interleukin 10
  • Interleukin 10 (IL-10) is the founding member of the IL-10 family of class II cytokines
  • All of the IL-10 cytokine family members have a four-helix bundle consisting of α-helical folds
  • Upon binding to its receptor, IL-10 activates signaling through JAK1 and STAT3
  • It is produced by dendritic cells, macrophages, and CD4+ T regulatory cells, as well as mast cells, NK cells, neutrophils, and regulatory B cells, under specific stimulating conditions
  • IL-10 can inhibit the activation of certain immune cells while it promotes the function of B cells, and facilitates the healing process
  • Specifically, this cytokine is important for the function of T regulatory cells as it is a potent suppressor of effector T cell proliferation and cytokine production
  • Also, IL-10 produced by a subset of macrophages inhibits activation and production of pro-inflammatory cytokines by neighboring macrophages, thus allowing a level of self-regulation
Specifications
Name B-TCGF, CSIF, Cytokine synthesis inhibitory factor, Interleukin-10, TGIF
Quantity 50 μg
Cell Type Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Myeloid Cells; Neurons; T Cells; T Cells, CD4+; T Cells, Regulatory
Purity or Quality Grade ≥95%