Learn More
Mouse Leptin R Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB497-SP
This item is not returnable.
View return policy
Description
Leptin R Monoclonal specifically detects Leptin R in Mouse samples. It is validated for Flow Cytometry, ELISA Capture (Matched Antibody Pair).
Specifications
| Leptin R | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3953 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry, ELISA | |
| 55305 | |
| Flow Cytometry 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P48356 | |
| LEPR | |
| Mouse myeloma cell line NS0-derived recombinant mouse Leptin R Leu21-Leu840 (predicted) Accession # P48356 | |
| 25 μg | |
| Primary | |
| Detects mouse Leptin R in ELISAs. | |
| Mouse | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu