missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-6, AOF Recombinant Protein

Product Code. 17636545
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 17636545

Brand: Invitrogen™ RP87728

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

E. coli Recombinant Protein

Interleukin 6 (IL-6) is a multifunctional 26 kD protein originally discovered in the medium of RNA-stimulated fibroblastoid cells. IL-6 appears to be directly involved in the responses that occur after infection and cellular injury, and it may prove to be important as IL-1 and TNF-a in regulating the acute phase response. IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages and endothelial cells. IL-6 acts upon a variety of cells including myeloid progenitor cells, T cells, B cells, and hepatocytes. In addition, IL-6 appears to interact with IL-2 in the proliferation of T lymphocytes. IL-6 potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors. IL-6 plays a critical role in B-cell differentiation to plasma cells and is a potent growth factor for plasmacytoma and myeloma. IL-6 is a very useful culture supplement for the generation of a high number of antibody-producing hybridomas. Primarily produced at sites of acute and chronic inflammation, IL-6 is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of IL-6 is implicated in a wide variety of inflammation-associated disease states including diabetes mellitus and systemic juvenile rheumatoid arthritis.
  • Purity: >95% by SDS-Page
  • Endotoxin Concentration: AX41 ≤EUs/μg
  • Protein Length: 188 aa.
  • Biological Activity: <50 pg/mL, <1 ng/mL
  • Molecular Weight: 21.9 kDa
  • Animal Origin Free: Yes
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08505
For Use With (Application) Control, ELISA, Functional Assay, Gel Shift, Western Blot
Formulation 0.1% TFA with no preservative
Gene ID (Entrez) 16193
Molecular Weight (g/mol) 21.9 kDa
Name Mouse IL-6, AOF
Quantity 100 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Gene Alias B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BOS_4106; BSF2; BSF-2; CDF; ChIL-6; CTL differentiation factor; EGM_12696; HGF; H-IL-6; HSF; Hybridoma growth factor; hypothetical protein; IFNB2; IFN-beta-2; IL6; Il-6; ILg6; ILN; Interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin BSF-2; Interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 protein; M-IL-6; R-IL-6
Gene Symbol Il6
Specific Reactivity Mouse
Conjugate Unconjugated
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.