missing translation for 'onlineSavingsMsg'
Learn More

Mouse IL-23R Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody

1875.00 NOK - 4440.00 NOK

Specifications

Antigen IL-23R
Clone 753317
Dilution Flow Cytometry 0.25 μg/106 cells, CyTOF-ready
Applications Flow Cytometry, CyTOF
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18780267
GREENER_CHOICE
View Documents
R&D Systems
MAB16861-SP
25 μg
1875.00 NOK
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18720707
GREENER_CHOICE
View Documents
R&D Systems
MAB16861-100
100 μg
4440.00 NOK
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-23R Monoclonal specifically detects IL-23R in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications

Specifications

IL-23R
Flow Cytometry 0.25 μg/106 cells, CyTOF-ready
Monoclonal
Supernatant
Mouse
Q8K4B4
149233.0
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
753317
Flow Cytometry, CyTOF
Unconjugated
Rat
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R, Gly24-Asp372, Accession No. Q8K4B4
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse IL-23 R in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.