missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IL-17A, AOF Recombinant Protein
Click to view available options
Quantity:
1 mg
100 μg
25 μg
500 μg
Unit Size:
100µg
1mg
25µg
500µg
Description
Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).
- Purity: >95% by SDS-Page
- Endotoxin Concentration: AX41 ≤EUs/μg
- Protein Length: 134 aa.
- Biological Activity: <5 ng/mL
- Molecular Weight: 15.1 kDa
- Animal Origin Free: Yes
Specifications
Specifications
| Accession Number | Q62386 |
| For Use With (Application) | Control, ELISA, Functional Assay, Gel Shift, Western Blot |
| Formulation | 10 mM sodium phosphate, 150 mM sodium sulfate with no preservative |
| Gene ID (Entrez) | 16171 |
| Molecular Weight (g/mol) | 15.1 kDa |
| Name | Mouse IL-17 A, AOF |
| Quantity | 1 mg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction