missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-17/IL-17A Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 17 publications
Brand: R&D Systems MAB721-SP
This item is not returnable.
View return policy
Description
IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| IL-17/IL-17A | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3605 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, ELISA | |
| 50101 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q62386 | |
| IL17A | |
| E. coli-derived recombinant mouse IL-17 Thr22-Ala158 Accession # Q62386 | |
| 25 μg | |
| Primary | |
| Detects mouse IL-17 in ELISAs and Western blots. In direct ELISAs, 100% reactivity with recombinant mouse (rm) IL-17A/IL-17F heterodimer, 50% cross-reactivity with recombinant human IL-17, and 25% cross-reactivity with recombinant canine IL-17 is observed. No cross-reactivity with rmIL-17B, rmIL-17C, rmIL-17D, rmIL-17E, or rmIL-17F is observed. In Western blots, cross-reactivity with recombinant rat IL-17A is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction