Learn More
Gibco™ Mouse IL-15 Recombinant Protein
Description
Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute mouse IL-15 in sterile, distilled water to a concentration of 0.5-1.0 mg/mL. Do not vortex. This solution can be stored at 2°C to 8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C. Storage: The lyophilized protein is stable at room temperature for 1 month, but should be kept at -20°C for long term storage. Working aliquots stored with a carrier protein are stable for at least 6 months at -20°C to -80°C. Avoid repeated freeze/thaw cycles.
Specifications
Specifications
| Accession Number | P48346 |
| For Use With (Application) | Bioactivity |
| Formulation | 10mM sodium phosphate with no preservative; pH 8 |
| Gene ID (Entrez) | 16168 |
| Molecular Weight (g/mol) | 13.4 kDa |
| Name | Mouse IL-15 |
| pH Range | 8 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Storage Requirements | -20°C |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.