missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-15/IL-15R Complex

Product Code. 15507506
Click to view available options
Quantity:
25 μg
5 μg
Unit Size:
25µg
5µg
This item is not returnable. View return policy

Product Code. 15507506

Brand: Invitrogen™ 14815262

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

Interleukin-15 (IL-15) is a 14 kDa proinflammatory protein that has been shown to play a role in the activation of neutrophils, dendritic cells, and macrophages, and is essential to the development and survival of NK cells and CD8 T-cells. IL-15 activates Jak/Stat pathways by signaling through a heterotrimeric receptor. Two of the subunits of this receptor, beta and gamma, are shared with the IL-2 receptor, while the alpha subunit is unique to IL-15.Despite the expression of IL-15 mRNA in many cell types, the secreted protein is rarely detectable in biological samples. Recent research suggests that IL-15 is retained inside the cell and is only secreted in complex with its unique receptor, IL-15R alpha. This chaperoning is required from the ER to Golgi through to secretion. The cytokine/receptor complex may stay bound to the cell surface, where it can be trans-presented to cells expressing the beta/gamma subunits of the receptor, or may be secreted in a soluble form that lacks the transmembrane domain of the receptor. The soluble IL-15/IL-15R complex is produced by mouse dendritic cells, fibroblasts, and macrophages, and demonstrates a 10-100-fold increase in agonistic activity over IL-15 alone. Description: This recombinant protein is the heterocomplex of the mouse IL-15 linked to soluble IL-15 receptor alpha chain (sushi domain) via a GS linker. Interleukin 15 (IL-15) is a cytokine that stimulates T and NK cell proliferation and differentiation. IL-15 regulates macrophage proinflammatory and chemotactic protein production. In addition, it induces differentiation of monocytes towards dendritic cells. The receptor for IL-15 is a high affinity heterotrimeric receptor composed of IL-15Ralpha and 2 additional subunits of the IL-2 receptor (beta and gamma). Upon activation, the receptor complex signals via the Stat and JAK pathways. It has been reported that the soluble form of IL-15Ralpha corresponding to the entire extracellular domain, cleaved by MMP′s, behaves as a high affinity IL-15 antagonist, whereas the recombinant extracellular domain comprised of the sushi domain together with the ligand, IL-15, behaves as a potent agonist by enhancing binding of IL-15 to the receptor complex. In fact, this receptor-ligand complex has approximately 10-100 fold increased proliferative activity. Applications Reported: Recombinant mouse IL-15/IL-15RA complex is biologically active. Applications Tested: The ED50 of this protein, as measured by CTLL-2 cell proliferation assay, is less than or equal to 4ng/mL. This corresponds to a specific activity of greater than or equal to 2.5 x 10e5 Units/mg. Source: Insect cell expressed N-terminal His-Tagged mIL-15Ra Gly33-Ala132 accession &Num; NP_032384-GS linker-mIL-15 Asn49-Ser162 accession &Num; NP_032383. Bioactivity: The ED50 of this protein, as measured by CTLL-2 cell proliferation assay, is less than or equal to 4ng/mL. This corresponds to a specific activity of greater than or equal to 2.5 x 10e5 Units/mg. Endotoxin: Less than 0.1ng/ug cytokine as determined by the LAL assay. Purity: >98% as determined by SDS-PAGE. Molecular Weight: 29.4 kDa. Storage and handling: For best recovery, quick-spin vial prior to opening. Use in a sterile environment. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2μm post-manufacturing filtered.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P48346, Q60819
Concentration 0.1mg/mL
For Use With (Application) Control, Functional Assay
Formulation TBS with 1% BSA and no preservative; pH 8.0
Gene ID (Entrez) 16168, 16169
Molecular Weight (g/mol) 29.4kDa
Name Mouse IL-15/IL-15R Complex
pH Range 8.0
Purification Method SDS-PAGE
Quantity 5 μg
Source Insect cells
Storage Requirements -80°C
Regulatory Status RUO
Structural Form Insect cell expressed N-terminal His-Tagged mIL-15Ra Gly33-Ala132 accession # NP_032384 - GS linker - mIL-15 Asn49-Ser162 accession # NP_032383
Endotoxin Concentration Less than 0.1ng/μg cytokine as determined by the LAL assay.
Common Name Mouse IL-15/IL-15R Complex
Gene Symbol Il15, Il15ra
Biological Activity The ED50 of this protein, as measured by CTLL-2 cell proliferation assay, is less than or equal to 4ng/mL. This corresponds to a specific activity of greater than or equal to 2.5 x 105 Units/mg.
Product Type Recombinant Protein
Specific Reactivity Mouse
Conjugate Unconjugated
Cross Reactivity Ms
Species Baculovirus
Recombinant Recombinant
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade Affinity chromatography
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.