missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IL-13 Recombinant Protein
E. coli Recombinant Protein
Brand: Invitrogen™ RP87883
Additional Details : Weight : 0.01000kg
Description
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.Specifications
P20109 | |
Control, ELISA, Western Blot | |
16163 | |
Mouse IL-13 (aa 26-131) | |
E. coli | |
RUO | |
ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; Interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600 | |
Mouse | |
Recombinant | |
>95% |
200 μg/mL | |
10 mM sodium phosphate with 150 mM NaCl and no preservative; pH 7.2 | |
11.7 kD | |
500 μg | |
-20°C | |
0.01 ng/ μg cytokine | |
Il13 | |
Unconjugated | |
Liquid |