Learn More
Mouse IgG2B APC-conjugated Antibody, R&D Systems™
Product Code. 18183701
Change view
Click to view available options
Quantity:
200 Tests
Unit Size:
200 tests
Product Code. Quantity unitSize
18183701 200 Tests 200 tests
1 options
Product Code. 18183701
Brand: R&D Systems IC0041A
Mouse Monoclonal Antibody has been used in 9 publications
IgG2b Monoclonal specifically detects IgG2b in Mouse samples. It is validated for Control.
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG2b
Classification Monoclonal
Clone 133303
Conjugate APC
Dilution Control 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Host Species Mouse
Immunogen KLH
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 200 Tests
Test Specificity Selected for use as mouse IgG 2B isotype control for flow cytometry applications.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2b