missing translation for 'onlineSavingsMsg'
Learn More

Mouse IGFALS/ALS Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18773379
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Product Code. Quantity unitSize
18773379 25 μg 25µg
1 options
This item is not returnable. View return policy

Product Code. 18773379

Brand: R&D Systems MAB1436SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

ALS Monoclonal specifically detects ALS in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALS
Applications Western Blot
Classification Monoclonal
Clone 199410
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P70389
Gene Alias ACLSD, ALS, Igfals
Gene Symbols Igfals
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IGFALS/ALS Thr28-Cys603 Accession # P70389
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3483
Test Specificity Detects mouse IGFALS/ALS in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.