missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse FLIP Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0013
Additional Details : Weight : 0.01000kg
Description
FLIP Synthetic Peptide, PEP-0013, from Invitrogen.
FLIP (CFLAR) is an apoptosis regulator protein which functions as a crucial link between cell survival and cell death pathways in mammalian cells. It acts as an inhibitor of TNFRSF6 mediated apoptosis. A proteolytic fragment (p43) is likely retained in the death-inducing signaling complex (DISC) thereby blocking further recruitment and processing of caspase-8 at the complex. Full length and shorter isoforms have been shown either to induce apoptosis or to reduce TNFRSF-triggered apoptosis. Lacks enzymatic (caspase) activity. This peptide corresponds to 17 amino acids near the carboxy terminus of mouse FLIP.PEP-0013 can be used as a blocking peptide with polyclonal antibody PA5-19881.Specifications
12633 | |
purified | |
Mouse FLIP | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Mouse | |
O35732 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
Cflar | |
0.02% sodium azide | |
50 μg | |
PBS |